Methyl-5-brom-2-methoxynicotinat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-bromo-2-methoxynicotinate, Methyl 5-bromo-2-methoxy-3-pyridinecarboxylate

OR3356-5G
APOSOR3356-5GEA 98.6 EUR
APOSOR3356-5G
Methyl-5-brom-2-methoxynicotinat 98%
Methyl-5-brom-2-methoxynicotinat
Formel: C₈H₈BrNO₃
Molecular Weight: 246,06 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08064227
CAS-Nummer: 122433-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR