2-[(Dimethylamino)methyl]-4-ethylbenzenol 95%
Lieferant: Apollo Scientific
OR33565-1G
OR33565-5G
APOSOR33565-1GEA
270
EUR
APOSOR33565-1G
APOSOR33565-5G
2-[(Dimethylamino)methyl]-4-ethylbenzenol 95%
2-[(Dimethylamino)methyl]-4-ethylbenzenol
Formel:
C₁₁H₁₇NO Molecular Weight: 179,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01734031 CAS-Nummer: 55955-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...