5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 97%
Lieferant: Apollo Scientific
OR33571-1G
OR33571-5G
APOSOR33571-1GEA
260
EUR
APOSOR33571-1G
APOSOR33571-5G
5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde 97%
5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Formel:
C₆H₄BrNO₂ Molecular Weight: 202,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09607921 CAS-Nummer: 952183-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...