tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme: 4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylic acid tert-Butyl ester, Ethyl [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetate, tert-Butyl 4-(2-Ethoxy-2-oxoethylidene)piperidine-1-carboxylate, (1-Boc-piperidin-4-ylidene)acetic acid ethyl ester, [1-(tert-Butoxycarbonyl)piperidin-4-ylidene]acetic acid ethyl ester, Ethyl (1-Boc-piperidin-4-ylidene)acetate
OR33587-1G OR33587-5G OR33587-25G
APOSOR33587-1GEA 53.8 EUR
APOSOR33587-1G APOSOR33587-5G APOSOR33587-25G
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate 95%
tert-Butyl-4-(2-ethoxy-2-oxoethylidene)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₄H₂₃NO₄
Molecular Weight: 269,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12912655
CAS-Nummer: 135716-08-4