1-[(2-Aminoethyl)sulphanyl]-4-bromobenzene 95%
Lieferant: Apollo Scientific
Synonyme:
2-[(4-Bromophenyl)sulphanyl]ethan-1-amine
OR33596-1G
APOSOR33596-1GEA
270
EUR
APOSOR33596-1G
1-[(2-Aminoethyl)sulphanyl]-4-bromobenzene 95%
1-[(2-Aminoethyl)sulphanyl]-4-bromobenzene
Formel:
C₈H₁₀BrNS Molecular Weight: 232,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05262934 CAS-Nummer: 69326-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...