2-[(4-Methoxyphenyl)amino]-2-methylpropanenitrile 97%

Lieferant: Apollo Scientific

OR33598-1G
APOSOR33598-1GEA 180 EUR
APOSOR33598-1G
2-[(4-Methoxyphenyl)amino]-2-methylpropanenitrile 97%
2-[(4-Methoxyphenyl)amino]-2-methylpropanenitrile
Formel: C₁₁H₁₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD06254727
CAS-Nummer: 893723-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR