Methyl-3-methyl-1-benzofuran-2-carboxylate 97%

Lieferant: Apollo Scientific

OR33601-1G
APOSOR33601-1GEA 435 EUR
APOSOR33601-1G
Methyl-3-methyl-1-benzofuran-2-carboxylate 97%
Methyl-3-methyl-1-benzofuran-2-carboxylate
Formel: C₁₁H₁₀O₃
Molecular Weight: 190,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09270117
CAS-Nummer: 2076-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR