6-(4-Methoxyphenyl)pyridine-3-carbaldehyde 97%

Lieferant: Apollo Scientific

OR33605-1G OR33605-5G
APOSOR33605-1GEA 336 EUR
APOSOR33605-1G APOSOR33605-5G
6-(4-Methoxyphenyl)pyridine-3-carbaldehyde 97%
6-(4-Methoxyphenyl)pyridine-3-carbaldehyde
Formel: C₁₃H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04115433
CAS-Nummer: 834884-62-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR