(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 95%
Lieferant: Apollo Scientific
Synonyme: N-(tert-Butoxycarbonyl)-D-proline methyll ester, Boc-D-Pro-OMe, N-Boc-D-proline methyll ester, N-Boc-D-proline methyl ester
OR33635-100G
APOSOR33635-100GEA 95.7 EUR
APOSOR33635-100G
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate 95%
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00190828
CAS-Nummer: 73323-65-6