2-Bromo-6-methoxy-1-indanone 95%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-6-methoxy-2,3-dihydro-1H-inden-1-one
OR33636-1G
APOSOR33636-1GEA
270
EUR
APOSOR33636-1G
2-Bromo-6-methoxy-1-indanone 95%
2-Bromo-6-methoxy-1-indanone
Formel:
C₁₀H₉BrO₂ Molecular Weight: 241,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02258470 CAS-Nummer: 62015-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...