2-Methyl-5-nitroindol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-5-nitro-1H-indole
OR3396-5G
OR3396-25G
APOSOR3396-5GEA
108
EUR
APOSOR3396-5G
APOSOR3396-25G
2-Methyl-5-nitroindol 95+%
2-Methyl-5-nitroindole
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,17 g/mol Schmelzpunkt: 176…178 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00090335 CAS-Nummer: 7570-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...