5-(Brommethyl)benzofurazan 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Bromomethyl)benzo[c][1,2,5]oxadiazole, 5-(Bromomethyl)-2,1,3-benzoxadiazole

OR3415-100MG OR3415-1G
APOSOR3415-100MGEA 29 EUR
APOSOR3415-100MG APOSOR3415-1G
5-(Brommethyl)benzofurazan 97%
5-(Brommethyl)benzofurazan
Formel: C₇H₅BrN₂O
Molecular Weight: 213,0314 g/mol
Schmelzpunkt: 77…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00174369
CAS-Nummer: 32863-31-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR