4-(1-Pyrrolylsulphonyl)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3432-1G
OR3432-500MG
OR3432-100MG
APOSOR3432-1GEA
261
EUR
APOSOR3432-1G
APOSOR3432-500MG
APOSOR3432-100MG
4-(1-Pyrrolylsulphonyl)benzeneboronic acid 98%
4-(1-Pyrrolylsulphonyl)benzeneboronic acid
Formel:
C₁₀H₁₀BNO₄S Molecular Weight: 251.063 g/mol Schmelzpunkt: 220…222 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689522 CAS-Nummer: 913835-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...