4-Cyano-3-iodopyridine 98%

Lieferant: Apollo Scientific
Achtung

OR345006-1G OR345006-250MG
APOSOR345006-1GEA 319 EUR
APOSOR345006-1G APOSOR345006-250MG
4-Cyano-3-iodopyridine 98%
4-Cyano-3-iodopyridine
Formel: C₆H₃IN₂
Molecular Weight: 230,01 g/mol
Schmelzpunkt: 119…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08729285
CAS-Nummer: 10386-28-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR