3,4-Dibrompyridin

Lieferant: Apollo Scientific
Achtung

OR345020-100G OR345020-25G OR345020-5G
APOSOR345020-100GEA 282 EUR
APOSOR345020-100G APOSOR345020-25G APOSOR345020-5G
3,4-Dibrompyridin
3,4-Dibromopyridine
Formel: C₅H₃Br₂N
Molecular Weight: 236,89 g/mol
Schmelzpunkt: 71…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234016
CAS-Nummer: 13534-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR