3-Amino-4-ethoxypyridine

Lieferant: Apollo Scientific

Synonyme: 4-Ethoxypyridin-3-amine

OR345032-1G OR345032-250MG OR345032-500MG
APOSOR345032-1GEA 84.1 EUR
APOSOR345032-1G APOSOR345032-250MG APOSOR345032-500MG
3-Amino-4-ethoxypyridine
3-Amino-4-ethoxypyridine
Formel: C₇H₁₀N₂O
Molecular Weight: 138.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234351
CAS-Nummer: 1633-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR