2-Methylnicotinonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Cyano-2-methylpyridine
OR345035-1G
OR345035-5G
APOSOR345035-1GEA
88.6
EUR
APOSOR345035-1G
APOSOR345035-5G
2-Methylnicotinonitrile
2-Methylnicotinonitrile
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06254420 CAS-Nummer: 1721-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...