[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%
Lieferant: Apollo Scientific
Synonyme:
2-(4-(Aminomethyl)phenoxy)-N,N-dimethylethanamine, 2-[4-(Amino methyl) phenoxy]-n, n-dimethyl ethanamine, Trimethobenzamide Impurity 1
OR345043-5G
OR345043-1G
APOSOR345043-5GEA
154
EUR
APOSOR345043-5G
APOSOR345043-1G
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine 97%
[2-(4-Aminomethylphenoxy)ethyl]dimethylamine
Formel:
C₁₁H₁₈N₂O Molecular Weight: 194.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075231 CAS-Nummer: 20059-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...