7-Chloro-1H-pyrrolo[2,3-c]pyridine 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Chloro-6-azaindole

OR345082-1G OR345082-5G
APOSOR345082-1GEA 85.7 EUR
APOSOR345082-1G APOSOR345082-5G
7-Chloro-1H-pyrrolo[2,3-c]pyridine 98%
7-Chloro-1H-pyrrolo[2,3-c]pyridine
Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03839905
CAS-Nummer: 357263-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR