2-Benzyloxy-5-hydroxypyridin 98%
Lieferant: Apollo Scientific
Synonyme:
6-(Benzyloxy)pyridin-3-ol
OR345132-5G
APOSOR345132-5GEA
145
EUR
APOSOR345132-5G
2-Benzyloxy-5-hydroxypyridin 98%
2-Benzyloxy-5-hydroxypyridin
Formel:
C₁₂H₁₁NO₂ Molecular Weight: 201,2212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798281 CAS-Nummer: 725256-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...