3-Hydroxy-5-nitrobenzoesäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR345140-1G
OR345140-25G
OR345140-5G
APOSOR345140-1GEA
58
EUR
APOSOR345140-1G
APOSOR345140-25G
APOSOR345140-5G
3-Hydroxy-5-nitrobenzoesäure 97%
3-Hydroxy-5-nitrobenzoic acid
Formel:
C₇H₅NO₅ Molecular Weight: 183,12 g/mol Schmelzpunkt: 190…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01098212 CAS-Nummer: 78238-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...