2-Benzyloxy-3-hydroxypyridin 95+%

Lieferant: Apollo Scientific
Achtung

OR345146-100MG OR345146-250MG
APOSOR345146-100MGEA 227 EUR
APOSOR345146-100MG APOSOR345146-250MG
2-Benzyloxy-3-hydroxypyridin 95+%
2-Benzyloxy-3-hydroxypyridin
Formel: C₁₂H₁₁NO₂
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06798282
CAS-Nummer: 885952-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR