4-Iod-2-methoxybenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR345152-1G
OR345152-25G
OR345152-5G
APOSOR345152-1GEA
21.9
EUR
APOSOR345152-1G
APOSOR345152-25G
APOSOR345152-5G
4-Iod-2-methoxybenzoesäure
4-Iodo-2-methoxybenzoic acid
Formel:
C₈H₇IO₃ Molecular Weight: 278,0439 g/mol Schmelzpunkt: 147…151 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07780726 CAS-Nummer: 89942-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...