3-Amino-2-brom-6-chlorpyridin 97%
Lieferant: Apollo Scientific
Synonyme:
2-Brom-6-chlor-3-pyridylamin
OR345185-1G
OR345185-5G
APOSOR345185-1GEA
0
EUR
APOSOR345185-1G
APOSOR345185-5G
3-Amino-2-brom-6-chlorpyridin 97%
3-Amino-2-bromo-6-chloropyridine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11504927 CAS-Nummer: 1050501-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...