8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin 96%
Lieferant: Apollo Scientific
OR345196-1G
OR345196-5G
APOSOR345196-1GEA
43.5
EUR
APOSOR345196-1G
APOSOR345196-5G
8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin 96%
8-Brom-6-chlor-3-nitroimidazo[1,2-a]pyridin
Formel:
C₇H₃BrClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11504898 CAS-Nummer: 1072944-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...