2,3-Dibrom-5-nitropyridin

Lieferant: Apollo Scientific

OR345271-5G
APOSOR345271-5GEA 68.3 EUR
APOSOR345271-5G
2,3-Dibrom-5-nitropyridin
2,3-Dibromo-5-nitropyridine
Formel: C₅H₂Br₂N₂O₂
Molecular Weight: 281,8896 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00233994
CAS-Nummer: 15862-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR