5-Acetyl-2-methoxypyridin 98%

Lieferant: Apollo Scientific

OR345295-25G
APOSOR345295-25GEA 237 EUR
APOSOR345295-25G
5-Acetyl-2-methoxypyridin 98%
5-Acetyl-2-methoxypyridin
Formel: C₈H₉NO₂
Molecular Weight: 151,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664350
CAS-Nummer: 213193-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR