2-Iodo-4-methylpyridine

Lieferant: Apollo Scientific

OR345296-1G OR345296-250MG OR345296-500MG
APOSOR345296-1GEA 458 EUR
APOSOR345296-1G APOSOR345296-250MG APOSOR345296-500MG
2-Iodo-4-methylpyridine
2-Iodo-4-methylpyridine
Formel: C₆H₆IN
Lagertemperatur: Raumtemperatur
MDL: MFCD12963767
CAS-Nummer: 22282-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR