2-Bromo-3-chloro-5-nitropyridine

Lieferant: Apollo Scientific

OR345300-5G
APOSOR345300-5GEA 184 EUR
APOSOR345300-5G
2-Bromo-3-chloro-5-nitropyridine
2-Bromo-3-chloro-5-nitropyridine
Formel: C₅H₂BrClN₂O₂
Molecular Weight: 237,4400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061302
CAS-Nummer: 22353-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR