6-Isopropylpyridin-3-amine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-2-isopropylpyridine

OR345336-1G OR345336-250MG
APOSOR345336-1GEA 104 EUR
APOSOR345336-1G APOSOR345336-250MG
6-Isopropylpyridin-3-amine 95%
6-Isopropylpyridin-3-amine
Formel: C₈H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08460245
CAS-Nummer: 405103-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR