2-Amino-5-chloropyridin-3-ol 98%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-5-chloro-3-hydroxypyridine, 2-Amino-3-hydroxy-5-chloropyridine
OR345338-25G
APOSOR345338-25GEA
94.4
EUR
APOSOR345338-25G
2-Amino-5-chloropyridin-3-ol 98%
2-Amino-5-chloropyridin-3-ol
Formel:
C₅H₅ClN₂O Molecular Weight: 144,5590 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706179 CAS-Nummer: 40966-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...