5-Bromo-3-chloropyridin-2(1H)-one 98%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-3-chloro-2-hydroxypyridine

OR345375-25G
APOSOR345375-25GEA 85.7 EUR
APOSOR345375-25G
5-Bromo-3-chloropyridin-2(1H)-one 98%
5-Bromo-3-chloropyridin-2(1H)-one
Formel: C₅H₃BrClNO
Molecular Weight: 208.44 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662389
CAS-Nummer: 58236-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR