3-Brom-4-methoxypyridin 98%

Lieferant: Apollo Scientific
Achtung

OR345416-100G
APOSOR345416-100GEA 150 EUR
APOSOR345416-100G
3-Brom-4-methoxypyridin 98%
3-Bromo-4-methoxypyridine
Formel: C₆H₆BrNO
Lagertemperatur: Gefrierschrank
MDL: MFCD05664024
CAS-Nummer: 82257-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR