2,3-Dichlor-6-methoxypyridin
Lieferant: Apollo Scientific
|
Gefahr
|
OR345420-1G
APOSOR345420-1GEA
56.7
EUR
APOSOR345420-1G
2,3-Dichlor-6-methoxypyridin
2,3-Dichloro-6-methoxypyridine
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,02 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08669402 CAS-Nummer: 83732-68-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...