3-Amino-6-chloropyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR345432-5G
APOSOR345432-5GEA 63.8 EUR
APOSOR345432-5G
3-Amino-6-chloropyridine-2-carboxylic acid
3-Amino-6-chloropyridine-2-carboxylic acid
Formel: C6H5ClN2O2
Molecular Weight: 172,5691 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264291
CAS-Nummer: 866807-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR