3-Brom-2-methoxybenzonitril 98%
Lieferant: Apollo Scientific
Synonyme:
3-Brom-2-methoxybenzoesäurenitril
OR345437-25G
APOSOR345437-25GEA
136
EUR
APOSOR345437-25G
3-Brom-2-methoxybenzonitril 98%
3-Brom-2-methoxybenzonitril
Formel:
C₈H₆BrNO Molecular Weight: 212,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834773 CAS-Nummer: 874472-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...