4-Chloro-5-iodopyridin-2-amine 98%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-4-chloro-5-iodopyridine
OR345478-5G
APOSOR345478-5GEA
62.5
EUR
APOSOR345478-5G
4-Chloro-5-iodopyridin-2-amine 98%
4-Chloro-5-iodopyridin-2-amine
Formel:
C₅H₄ClIN₂ Molecular Weight: 254,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977433 CAS-Nummer: 670253-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...