4-Amino-5-nitro-2-pyridinol 95%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-2-hydroxy-5-nitropyridine
OR345542-1G
OR345542-250MG
APOSOR345542-1GEA
246
EUR
APOSOR345542-1G
APOSOR345542-250MG
4-Amino-5-nitro-2-pyridinol 95%
4-Amino-5-nitro-2-pyridinol
Formel:
C₅H₅N₃O₃ Molecular Weight: 155,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040284 CAS-Nummer: 99479-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...