4-Chlor-2-methyl-3-nitropyridin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlor-3-nitro-2-picolin

OR345551-1G OR345551-250MG
APOSOR345551-1GEA 150 EUR
APOSOR345551-1G APOSOR345551-250MG
4-Chlor-2-methyl-3-nitropyridin 95%
4-Chloro-2-methyl-3-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215572
CAS-Nummer: 23056-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR