5,6-Dibromopyridin-3-amine 95+%
Lieferant: Apollo Scientific
Synonyme:
5-Amino-2,3-dibromopyridine
OR345555-1G
OR345555-5G
APOSOR345555-1GEA
52.2
EUR
APOSOR345555-1G
APOSOR345555-5G
5,6-Dibromopyridin-3-amine 95+%
5,6-Dibromopyridin-3-amine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,9100 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110698 CAS-Nummer: 53242-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...