6-Bromo-2-chloro-3-iodopyridine 97%

Lieferant: Apollo Scientific
Gefahr

OR345567-5G OR345567-1G OR345567-25G
APOSOR345567-5GEA 125 EUR
APOSOR345567-5G APOSOR345567-1G APOSOR345567-25G
6-Bromo-2-chloro-3-iodopyridine 97%
6-Bromo-2-chloro-3-iodopyridine
Formel: C₅H₂BrClIN
Lagertemperatur: Raumtemperatur
MDL: MFCD11857690
CAS-Nummer: 1138444-17-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR