N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester

Lieferant: Apollo Scientific

OR346005-1G OR346005-500MG
APOSOR346005-1GEA 0 EUR
APOSOR346005-1G APOSOR346005-500MG
N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-(4-methoxyphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Formel: C₁₃H₁₉N₃O₃
Molecular Weight: 265,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568181
CAS-Nummer: 1053655-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR