N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Lieferant: Apollo Scientific

OR346015-1G OR346015-500MG
APOSOR346015-1GEA 0 EUR
APOSOR346015-1G APOSOR346015-500MG
N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(2-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formel: C₁₃H₁₈ClN₃O₂
Molecular Weight: 283,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568195
CAS-Nummer: 1053656-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR