N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester

Lieferant: Apollo Scientific

OR346043-1G OR346043-500MG
APOSOR346043-1GEA 0 EUR
APOSOR346043-1G APOSOR346043-500MG
N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclobutylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Formel: C₁₇H₂₅N₃O₂
Molecular Weight: 303.41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568257
CAS-Nummer: 1053657-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR