6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester

Lieferant: Apollo Scientific
Achtung

OR346110-1G OR346110-500MG
APOSOR346110-1GEA 0 EUR
APOSOR346110-1G APOSOR346110-500MG
6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester
6H-Furo[2,3-b]pyrrole-5-carboxylic acid methyl ester
Formel: C₈H₇NO₃
Molecular Weight: 165.15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11052464
CAS-Nummer: 201019-27-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR