4H-Furo[3,2-b]pyrrole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR346111-1G
OR346111-500MG
APOSOR346111-1GEA
0
EUR
APOSOR346111-1G
APOSOR346111-500MG
4H-Furo[3,2-b]pyrrole-5-carboxylic acid
4H-Furo[3,2-b]pyrrole-5-carboxylic acid
Formel:
C₇H₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368660 CAS-Nummer: 67268-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...