4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde
Lieferant: Apollo Scientific
OR346115-25G
APOSOR346115-25GEA
190
EUR
APOSOR346115-25G
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde
4,6-Dichloro-2-(methylthio)pyrimidine-5-carbaldehyde
Formel:
C₆H₄Cl₂N₂OS Molecular Weight: 223,08 g/mol Schmelzpunkt: 86…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00194954 CAS-Nummer: 33097-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...