5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester
Lieferant: Apollo Scientific
OR346116-1G
OR346116-5G
APOSOR346116-1GEA
0
EUR
APOSOR346116-1G
APOSOR346116-5G
5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester
5-Dimethoxymethyl-1-phenyl-1H-pyrazole-4-carboxylic acid methyl ester
Formel:
C₁₄H₁₆N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02090518 CAS-Nummer: 957947-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...