4-Nitro-1H-pyrazole-3,5-dicarboxylic acid

Lieferant: Apollo Scientific

OR346122-1G OR346122-5G
APOSOR346122-1GEA 0 EUR
APOSOR346122-1G APOSOR346122-5G
4-Nitro-1H-pyrazole-3,5-dicarboxylic acid
4-Nitro-1H-pyrazole-3,5-dicarboxylic acid
Formel: C₅H₃N₃O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD01076649
CAS-Nummer: 62078-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR