1-tert-Butyl-5-methyl-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
Synonyme: 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carboxylic acid
OR346124-1G OR346124-5G
APOSOR346124-1GEA 217 EUR
APOSOR346124-1G APOSOR346124-5G
1-tert-Butyl-5-methyl-1H-pyrazole-3-carboxylic acid
1-tert-Butyl-5-methyl-1H-pyrazole-3-carboxylic acid
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01313628
CAS-Nummer: 376387-68-7